Gene Transcripts
- Gene Symbol
- Vwa5b2
- Entrez Gene ID
- 328643
- Gene Name
- von Willebrand factor A domain containing 5B2
- Gene Aliases
- BB182430, EG328644, Gm1738, Gm931
- Chromosome Location
- Chr. 16: 20589319 - 20605378 on Build GRCm38
- UniGene ID
- Mm.40539
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK045595.1 |
- |
1 |
1364 |
|
BC110635.2 |
- |
1 |
1103 |
|
BC125016.1 |
- |
1 |
1105 |
|
View Full Product Details