Gene Transcripts

Gene Symbol
A4gnt
Entrez Gene ID
333424
Gene Name
alpha-1,4-N-acetylglucosaminyltransferase
Gene Aliases
AV080780, Alpha4gnt, Gm798
Chromosome Location
Chr. 9: 99591175 - 99622367 on Build GRCm38
UniGene ID
Mm.297753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077424.2 NP_001070892.1 2 338
GenBank mRNA AK136425.1 - 2 340
BC115614.1 - 1 229 40102739
BC115615.1 - 1 229 40102743

View Full Product Details