Gene Transcripts

Gene Symbol
Gm13889
Entrez Gene ID
620695
Gene Name
predicted gene 13889
Gene Aliases
Ag2, OTTMUSG00000014994
Chromosome Location
Chr. 2: 93955810 - 93957100 on Build GRCm38
UniGene ID
Mm.482106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145034.1 NP_001138506.1 1 529
GenBank mRNA AK143447.1 - 1 478

View Full Product Details