Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026688.2 | NP_080964.1 | 4 | 437 | |
GenBank mRNA | AK002501.1 | - | 4 | 410 | |
AK088337.1 | - | 4 | 438 | ||
AK160888.1 | - | 2 | 2200 | ||
AK185633.1 | - | 2 | 219 | ||
AK206263.1 | - | 2 | 219 | ||
BC027270.1 | - | 4 | 391 | 3491580 | |
BC119267.1 | - | 4 | 391 | 8734016 | |
BC119269.1 | - | 4 | 392 | 8734018 |