Gene Transcripts

Gene Symbol
SECTM1
Entrez Gene ID
6398
Gene Name
secreted and transmembrane 1
Gene Aliases
K12
Chromosome Location
Chr. 17: 82321024 - 82334045 on Build GRCh38
UniGene ID
Hs.558009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003004.2 NP_002995.1 3 555
XM_005256392.3 XP_005256449.1 3 339
XM_011523588.2 XP_011521890.1 3 586
GenBank mRNA AK223068.1 - 3 248
AK223104.1 - 3 279
AK312980.1 - 3 275
BC017716.1 - 3 249 4413387
U77643.1 - 3 275

View Full Product Details