Gene Transcripts
- Gene Symbol
- Nrxn1
- Entrez Gene ID
- 60391
- Gene Name
- neurexin 1
- Gene Aliases
- Nrxn1b
- Chromosome Location
- Chr. 6: 13886757 - 13908139 on Build Rnor_6.0
- UniGene ID
- Rn.54924
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021767.2 |
NP_068535.2
|
|
1777 |
|
GenBank mRNA |
M96374.1 |
- |
|
1777 |
|
View Full Product Details