Gene Transcripts
- Gene Symbol
- Akp3
- Entrez Gene ID
- 64621
- Gene Name
- alkaline phosphatase 3, intestine, not Mn requiring
- Gene Aliases
- Alpi2, IAP-2, IAP-II
- Chromosome Location
- Chr. 9: 94228960 - 94232001 on Build Rnor_6.0
- UniGene ID
- Rn.138122
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022680.1 |
NP_073171.1
|
5, 6 |
647 |
|
GenBank mRNA |
S66545.1 |
- |
|
700 |
|
View Full Product Details