Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006755.1 | NP_006746.1 | 4 | 462 | |
GenBank mRNA | AK295537.1 | - | 4 | 430 | |
AK313427.1 | - | 4 | 430 | ||
BC010103.2 | - | 4 | 467 | 3536661 | |
BC018847.2 | - | 4 | 410 | ||
L19437.2 | - | 4 | 462 |