Gene Transcripts

Gene Symbol
Fndc5
Entrez Gene ID
260327
Gene Name
fibronectin type III domain containing 5
Gene Aliases
Chromosome Location
Chr. 5: 147323240 - 147330266 on Build Rnor_6.0
UniGene ID
Rn.18757
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270981.1 NP_001257910.1 5 581
GenBank mRNA AF529211.1 - 4 487

View Full Product Details