Gene Transcripts

Gene Symbol
B3gnt6
Entrez Gene ID
292325
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase)
Gene Aliases
RGD1310926
Chromosome Location
Chr. 1: 163143027 - 163149495 on Build Rnor_6.0
UniGene ID
Rn.104873
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106211.1 NP_001099681.1 2 587
XM_006229745.2 XP_006229807.1 2 1142

View Full Product Details