Gene Transcripts

Gene Symbol
B3gnt1
Entrez Gene ID
293667
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1
Gene Aliases
B3gnt6
Chromosome Location
Chr. 1: 220416018 - 220418240 on Build Rnor_6.0
UniGene ID
Rn.219389
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106324.1 NP_001099794.1 703

Gene Symbol
LOC100911750
Entrez Gene ID
100911750
Gene Name
N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase-like
Gene Aliases
Chromosome Location
Chr. 1: 220322811 - 220325078 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749036.3 XP_003749084.1 1 746

View Full Product Details