Gene Transcripts
- Gene Symbol
- Cpg1
- Entrez Gene ID
- 299247
- Gene Name
- candidate plasticity gene 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 123034304 - 123046297 on Build Rnor_6.0
- UniGene ID
- Rn.10347
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178104.2 |
NP_835205.1
|
2 |
890 |
|
GenBank mRNA |
X73292.1 |
- |
2 |
890 |
|
View Full Product Details