Gene Transcripts

Gene Symbol
AQP7P5
Entrez Gene ID
102724655
Gene Name
aquaporin 7 pseudogene 5
Gene Aliases
Chromosome Location
Chr. 9: 40463754 - 40499011 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017015366.1 XP_016870855.1 9 1333
XM_017015367.1 XP_016870856.1 8 1250
XM_017015368.1 XP_016870857.1 8 1233
XM_017015369.1 XP_016870858.1 9 1316
XM_017015370.1 XP_016870859.1 5 1333
XM_017015371.1 XP_016870860.1 7 1100
XM_017015372.1 XP_016870861.1 6 1412
XM_017015373.1 XP_016870862.1 6 971
XM_017015374.1 XP_016870863.1 6 909
XM_017015375.1 XP_016870864.1 5 825

Gene Symbol
AQP7
Entrez Gene ID
364
Gene Name
aquaporin 7
Gene Aliases
AQP7L, AQPap, GLYCQTL
Chromosome Location
Chr. 9: 33383135 - 33402682 on Build GRCh38
UniGene ID
Hs.455323
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170.2 NP_001161.1 8 1233
NM_001318156.1 NP_001305085.1 7 1056
NM_001318157.1 NP_001305086.1 7 1909
NM_001318158.1 NP_001305087.1 7 1591
NR_134513.1 5 1348
NR_134514.1 6 1467
NR_134515.1 7 1618
XM_005251453.3 XP_005251510.1 9 1333
XM_006716765.3 XP_006716828.1 7 994
XM_011517866.2 XP_011516168.1 8 1143
XM_011517867.2 XP_011516169.1 6 773
XM_017014700.1 XP_016870189.1 8 1434
XM_017014701.1 XP_016870190.1 7 1104
XM_017014702.1 XP_016870191.1 8 1139
XM_017014703.1 XP_016870192.1 7 892
XM_017014704.1 XP_016870193.1 6 1200
XM_017014705.1 XP_016870194.1 6 870
XM_017014706.1 XP_016870195.1 8 1622
GenBank mRNA AB006190.1 - 8 1070
AK055744.1 - 1 1175
BC062701.1 - 7 883 30324263
BC119672.1 - 6 1299
BC119673.1 - 6 1299
DQ786216.1 - 3 445

Gene Symbol
AQP7P1
Entrez Gene ID
375719
Gene Name
aquaporin 7 pseudogene 1
Gene Aliases
bA251O17.3
Chromosome Location
Chr. 9: 63315243 - 63334520 on Build GRCh38
UniGene ID
Hs.658215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002817.2 4 1226
GenBank mRNA AK128231.1 - 4 1222

Gene Symbol
LOC100509620
Entrez Gene ID
100509620
Gene Name
putative aquaporin-7-like protein 3
Gene Aliases
Chromosome Location
Chr. 2: 94587240 - 94613324 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005276965.4 XP_005277022.1 5 724
XM_006712884.3 XP_006712947.1 5 1148
XM_006712887.3 XP_006712950.1 7 1101
XM_017005467.1 XP_016860956.1 4 720
XM_017005468.1 XP_016860957.1 4 628
XM_017005469.1 XP_016860958.1 4 617

View Full Product Details