Gene Transcripts

Gene Symbol
IL1R2
Entrez Gene ID
7850
Gene Name
interleukin 1 receptor type 2
Gene Aliases
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
Chromosome Location
Chr. 2: 101991805 - 102028544 on Build GRCh38
UniGene ID
Hs.25333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261419.1 NP_001248348.1 4 716
NM_004633.3 NP_004624.1 4 716
NR_048564.1 4 703
XM_006712734.3 XP_006712797.1 4 740
XM_006712736.3 XP_006712799.1 4 777
XM_011511801.2 XP_011510103.1 4 3076
XM_011511803.2 XP_011510105.1 4 2655
XM_011511804.2 XP_011510106.1 4 769
XM_011511805.2 XP_011510107.1 4 1542
XM_011511807.1 XP_011510109.1 4 3076
XM_017004889.1 XP_016860378.1 3 626
GenBank mRNA AK130412.1 - 4 600
AK309161.1 - 3 472
BC027733.2 - 4 670
BC039031.1 - 4 716 5723005
FJ200480.1 - 3 327
U64094.1 - 3 548
U74649.1 - 3 522
X59770.1 - 3 548

View Full Product Details