Gene Transcripts

Gene Symbol
Rundc1
Entrez Gene ID
303552
Gene Name
RUN domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 89338991 - 89349230 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750935.3 XP_003750983.1 5 1203
XM_006247467.2 XP_006247529.1 6 1209

View Full Product Details