Gene Transcripts
- Gene Symbol
- SHOC2
- Entrez Gene ID
- 8036
- Gene Name
- SHOC2, leucine rich repeat scaffold protein
- Gene Aliases
- SIAA0862, SOC2, SUR8
- Chromosome Location
- Chr. 10: 110919370 - 111013667 on Build GRCh38
- UniGene ID
- Hs.104315
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001269039.2 |
NP_001255968.1
|
4, 5 |
1380 |
|
NM_001324336.1 |
NP_001311265.1
|
5, 6 |
1475 |
|
NM_001324337.1 |
NP_001311266.1
|
6, 7 |
1703 |
|
NM_007373.3 |
NP_031399.2
|
5, 6 |
1518 |
|
NR_136749.1 |
|
4, 5 |
581 |
|
XM_017016702.1 |
XP_016872191.1
|
5, 6 |
1423 |
|
XM_017016703.1 |
XP_016872192.1
|
5, 6 |
7240 |
|
XM_017016704.1 |
XP_016872193.1
|
5, 6 |
1422 |
|
GenBank mRNA |
AB020669.1 |
- |
5, 6 |
1446 |
|
AF054828.1 |
- |
5, 6 |
1440 |
|
AF068920.1 |
- |
4, 5 |
1169 |
|
AK090820.1 |
- |
3, 4 |
539 |
|
AK292833.1 |
- |
6, 7 |
1644 |
|
AK293271.1 |
- |
4, 5 |
520 |
|
BC044752.1 |
- |
5, 6 |
1454 |
|
BC050445.1 |
- |
4, 5 |
1373 |
6067669 |
BX649055.1 |
- |
6, 7 |
1286 |
|
View Full Product Details