Gene Transcripts

Gene Symbol
C1qtnf7
Entrez Gene ID
305423
Gene Name
C1q and tumor necrosis factor related protein 7
Gene Aliases
Ctrp7
Chromosome Location
Chr. 14: 72011518 - 72122264 on Build Rnor_6.0
UniGene ID
Rn.35545
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107221.1 NP_001100691.1 2, 3 310
XM_006251073.2 XP_006251135.1 2, 3 868
XM_006251075.2 XP_006251137.1 2, 3 1571
XM_008770208.1 XP_008768430.1 1, 2 808
GenBank mRNA BC166720.1 - 2, 3 698
FQ220553.1 - 2, 3 308

View Full Product Details