Gene Transcripts

Gene Symbol
Rgma
Entrez Gene ID
308739
Gene Name
repulsive guidance molecule family member A
Gene Aliases
Chromosome Location
Chr. 1: 134699053 - 134742512 on Build Rnor_6.0
UniGene ID
Rn.22326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107524.1 NP_001100994.1 4 1202
XM_008759521.1 XP_008757743.1 2 2649

View Full Product Details