Gene Transcripts

Gene Symbol
Adh6
Entrez Gene ID
310903
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
Chromosome Location
Chr. 2: 243656566 - 243687857 on Build Rnor_6.0
UniGene ID
Rn.214994
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012084.1 NP_001012084.1 8 1075
XM_006233372.2 XP_006233434.1 8 1869
XM_006233374.1 XP_006233436.1 9 1147
XM_006233375.2 XP_006233437.1 10 1358
XM_006233377.2 XP_006233439.1 8 1317
XM_006233378.2 XP_006233440.1 9 1227
XM_006233379.2 XP_006233441.1 9 1224
XM_008761506.1 XP_008759728.1 7 1189
XM_008761507.1 XP_008759729.1 9 3421
XM_008761508.1 XP_008759730.1 10 3281
XM_008761509.1 XP_008759731.1 10 1773
XM_008761510.1 XP_008759732.1 10 3549
GenBank mRNA BC083792.1 - 8 1075

View Full Product Details