Gene Transcripts

Gene Symbol
Amotl1
Entrez Gene ID
315430
Gene Name
angiomotin-like 1
Gene Aliases
Chromosome Location
Chr. 8: 13037265 - 13156989 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766026.1 XP_008764248.1 14 2635

View Full Product Details