Gene Transcripts

Gene Symbol
ASMTL
Entrez Gene ID
8623
Gene Name
acetylserotonin O-methyltransferase-like
Gene Aliases
ASMTLX, ASMTLY, ASTML
Chromosome Location
Chr. X: 1403139 - 1453794 on Build GRCh38
UniGene ID
Hs.533514
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173473.1 NP_001166944.1 4, 5 396
NM_001173474.1 NP_001166945.1 3, 4 433
NM_004192.3 NP_004183.2 4, 5 481
XM_005274434.3 XP_005274491.1 5, 6 587
XM_011546185.2 XP_011544487.1 2, 3 229
XM_017029919.1 XP_016885408.1 3, 4 277
GenBank mRNA AK001090.1 - 4, 5 377
AK090498.1 - 3, 4 385
AK225770.1 - 4, 5 403
AK299231.1 - 4, 5 399
AK301844.1 - 4, 5 390
BC002508.2 - 4, 5 367 3051194
BC010089.2 - 4, 5 365 2988425
DA005122.1 - 4, 5 481
DA301876.1 - 4, 5 436
DA904467.1 - 4, 5 396

View Full Product Details