Gene Transcripts

Gene Symbol
Arx
Entrez Gene ID
317268
Gene Name
aristaless related homeobox
Gene Aliases
RGD1562672
Chromosome Location
Chr. X: 62363757 - 62376139 on Build Rnor_6.0
UniGene ID
Rn.22484
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100174.1 NP_001093644.1 6 2455
GenBank mRNA EF638804.1 - 6 2455

View Full Product Details