Gene Transcripts

Gene Symbol
Tmem17
Entrez Gene ID
360985
Gene Name
transmembrane protein 17
Gene Aliases
Chromosome Location
Chr. 14: 107302808 - 107308226 on Build Rnor_6.0
UniGene ID
Rn.8102
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010961.1 NP_001010961.1 4 712
GenBank mRNA BC089059.1 - 4 712
FQ228748.1 - 4 370

View Full Product Details