Gene Transcripts

Gene Symbol
Tmem87b
Entrez Gene ID
362212
Gene Name
transmembrane protein 87B
Gene Aliases
RGD1307907
Chromosome Location
Chr. 3: 121357802 - 121393443 on Build Rnor_6.0
UniGene ID
Rn.28728
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191854.1 NP_001178783.1 19 1636
XM_006234976.2 XP_006235038.1 19 1840

View Full Product Details