Gene Transcripts

Gene Symbol
Gpsm3
Entrez Gene ID
406163
Gene Name
G-protein signaling modulator 3
Gene Aliases
AGS4, G18, Ng1
Chromosome Location
Chr. 20: 4355246 - 4357107 on Build Rnor_6.0
UniGene ID
Rn.7475
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003974.2 NP_001003974.1 4 950
GenBank mRNA BC088220.1 - 4 950
FQ229559.1 - 4 770

View Full Product Details