Gene Transcripts

Gene Symbol
Tryx5
Entrez Gene ID
408205
Gene Name
trypsin X5
Gene Aliases
Chromosome Location
Chr. 4: 69156919 - 69164449 on Build Rnor_6.0
UniGene ID
Rn.143124
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009173.1 NP_001009173.1 5 700
XM_006236368.2 XP_006236430.1 5 1271

View Full Product Details