Gene Transcripts

Gene Symbol
Man1b1
Entrez Gene ID
499751
Gene Name
mannosidase, alpha, class 1B, member 1
Gene Aliases
ERMan1, RGD1563595
Chromosome Location
Chr. 3: 2547986 - 2569050 on Build Rnor_6.0
UniGene ID
Rn.86273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109196.1 NP_001102666.1 4 480
GenBank mRNA BC166449.1 - 4 480
FQ210932.1 - 1 60

View Full Product Details