Gene Transcripts

Gene Symbol
Ctag2
Entrez Gene ID
501655
Gene Name
cancer/testis antigen 2
Gene Aliases
RGD1561702
Chromosome Location
Chr. X: 148421627 - 148422928 on Build Rnor_6.0
UniGene ID
Rn.126531
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109322.1 NP_001102792.1 1 142
XM_008773587.1 XP_008771809.1 1 124

View Full Product Details