Gene Transcripts

Gene Symbol
Hist1h1b
Entrez Gene ID
680522
Gene Name
histone cluster 1, H1b
Gene Aliases
Chromosome Location
Chr. 17: 44766313 - 44766981 on Build Rnor_6.0
UniGene ID
Rn.218561
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109417.1 NP_001102887.1 1 247

View Full Product Details