Gene Transcripts

Gene Symbol
Ccdc28b
Entrez Gene ID
682445
Gene Name
coiled coil domain containing 28B
Gene Aliases
Chromosome Location
Chr. 5: 147823446 - 147830430 on Build Rnor_6.0
UniGene ID
Rn.216577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134689.1 NP_001128161.1 6 724
XM_008764171.1 XP_008762393.1 6 3519
GenBank mRNA BC168155.1 - 5 1101

View Full Product Details