Gene Transcripts

Gene Symbol
Sssca1
Entrez Gene ID
689397
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
Chromosome Location
Chr. 1: 221087803 - 221089586 on Build Rnor_6.0
UniGene ID
Rn.6896
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109537.2 NP_001103007.1 2 152

View Full Product Details