Gene Transcripts

Gene Symbol
Tvp23b
Entrez Gene ID
687358
Gene Name
trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
Gene Aliases
Fam18b2
Chromosome Location
Chr. 10: 49299125 - 49310402 on Build Rnor_6.0
UniGene ID
Rn.75470
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107568.3 NP_001101038.2 5 407
GenBank mRNA BC168178.1 - 5 405
CK483802.1 - 5 407
FQ212667.1 - 5 443
FQ219631.1 - 5 408

View Full Product Details