Gene Transcripts

Gene Symbol
Dcun1d2
Entrez Gene ID
688913
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 2
Gene Aliases
RGD1311743
Chromosome Location
Chr. 16: 81008177 - 81034954 on Build Rnor_6.0
UniGene ID
Rn.15476
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134798.1 NP_001128270.1 6 683
GenBank mRNA FQ224263.1 - 6 696

Gene Symbol
NEWGENE_1582994
Entrez Gene ID
103694000
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 2
Gene Aliases
Dcun1d2
Chromosome Location
Chr. 16: 81587292 - 81614544 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771413.1 XP_008769635.1 6 851

View Full Product Details