Gene Transcripts

Gene Symbol
IQSEC1
Entrez Gene ID
9922
Gene Name
IQ motif and Sec7 domain 1
Gene Aliases
ARF-GEP100, ARFGEP100, BRAG2, GEP100
Chromosome Location
Chr. 3: 12897043 - 13282998 on Build GRCh38
UniGene ID
Hs.475506
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134382.2 NP_001127854.1 3 575
NM_014869.6 NP_055684.3 3 761
XM_011534304.2 XP_011532606.1 5 812
XM_011534305.2 XP_011532607.1 3 561
XM_011534306.2 XP_011532608.1 3 967
XM_011534307.2 XP_011532609.1 4 913
XM_011534308.2 XP_011532610.1 3 756
XM_011534310.2 XP_011532612.1 5 812
XM_011534311.2 XP_011532613.1 3 924
XM_011534312.2 XP_011532614.1 3 552
XM_011534313.2 XP_011532615.1 3 771
XM_011534314.2 XP_011532616.1 3 507
XM_011534315.2 XP_011532617.1 3 510
XM_017007581.1 XP_016863070.1 5 812
XM_017007582.1 XP_016863071.1 4 767
GenBank mRNA AB018306.1 - 2 254
AK294525.1 - 3 575
AY653734.1 - 3 523
BC010267.1 - 3 515 2900367

View Full Product Details