Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039659.1 | NP_001034748.1 | 4 | 453 | |
NM_001039660.1 | NP_001034749.1 | 3 | 341 | ||
NM_001145055.1 | NP_001138527.1 | 2 | 233 | ||
NM_001145057.1 | NP_001138529.1 | 3 | 279 | ||
NM_005699.3 | NP_005690.2 | 2 | 1097 | ||
NM_173042.2 | NP_766630.2 | 2 | 1097 | ||
NM_173044.2 | NP_766632.2 | 3 | 341 | ||
XM_017017059.1 | XP_016872548.1 | 3 | 1043 | ||
XM_017017060.1 | XP_016872549.1 | 3 | 1010 | ||
XM_017017061.1 | XP_016872550.1 | 4 | 906 | ||
XM_017017062.1 | XP_016872551.1 | 4 | 779 | ||
XM_017017063.1 | XP_016872552.1 | 4 | 1355 | ||
GenBank mRNA | AB019504.1 | - | 2 | 175 | |
AF110799.1 | - | 2 | 234 | ||
AF110800.1 | - | 2 | 234 | ||
AF110801.1 | - | 2 | 1097 | ||
AF122906.1 | - | 3 | 253 | ||
AF215907.2 | - | 3 | 330 | ||
AK098255.1 | - | 2 | 2191 | ||
BC044215.1 | - | 4 | 435 | 5755934 | |
BX357194.2 | - | 4 | 473 | ||
CB154498.1 | - | 3 | 344 |