Gene Transcripts
- Gene Symbol
- LOC100288884
- Entrez Gene ID
- 100288884
- Gene Name
- uncharacterized LOC100288884
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.613108
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124122.1 |
- |
2 |
343 |
|
- Gene Symbol
- C2orf27A
- Entrez Gene ID
- 29798
- Gene Name
- chromosome 2 open reading frame 27A
- Gene Aliases
- C2orf27, C2orf27B
- Chromosome Location
- Chr. 2: 131722491 - 131767404 on Build GRCh38
- UniGene ID
- Hs.635289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309649.1 |
- |
1 |
424 |
|
- Gene Symbol
- LOC105372731
- Entrez Gene ID
- 105372731
- Gene Name
- CMT1A duplicated region transcript 15 protein-like protein
- Gene Aliases
- Chromosome Location
- Chr. 21: 9026613 - 9036543 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details