Gene Transcripts
- Gene Symbol
- PATJ
- Entrez Gene ID
- 10207
- Gene Name
- PATJ, crumbs cell polarity complex component
- Gene Aliases
- Cipp, INADL, InaD-like, hINADL
- Chromosome Location
- Chr. 1: 61742477 - 62163920 on Build GRCh38
- UniGene ID
- Hs.478125
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_176877.2 |
NP_795352.2
|
35, 36 |
4762 |
|
XM_005270341.3 |
XP_005270398.1
|
35, 36 |
4760 |
|
XM_011540462.2 |
XP_011538764.1
|
37, 38 |
4925 |
|
XM_011540463.2 |
XP_011538765.1
|
37, 38 |
5113 |
|
XM_011540464.2 |
XP_011538766.1
|
37, 38 |
4877 |
|
XM_011540465.2 |
XP_011538767.1
|
36, 37 |
4850 |
|
XM_011540466.2 |
XP_011538768.1
|
36, 37 |
4835 |
|
XM_011540467.2 |
XP_011538769.1
|
37, 38 |
4925 |
|
XM_016999998.1 |
XP_016855487.1
|
35, 36 |
4751 |
|
XM_016999999.1 |
XP_016855488.1
|
34, 35 |
4676 |
|
XM_017000000.1 |
XP_016855489.1
|
32, 33 |
4171 |
|
XM_017000001.1 |
XP_016855490.1
|
18, 19 |
2002 |
|
GenBank mRNA |
AB044807.1 |
- |
22, 23 |
3041 |
|
AF397170.1 |
- |
34, 35 |
4648 |
|
AK293519.1 |
- |
10, 11 |
1168 |
|
AK308665.1 |
- |
9, 10 |
1325 |
|
BC021135.1 |
- |
2, 3 |
283 |
|
View Full Product Details