Gene Transcripts

Gene Symbol
Mtpn
Entrez Gene ID
14489
Gene Name
myotrophin
Gene Aliases
5033418D15Rik, Gcdp, V1
Chromosome Location
Chr. 6: 35508824 - 35539888 on Build GRCm38
UniGene ID
Mm.182746
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008098.4 NP_032124.1 3, 4 549
GenBank mRNA AK002620.1 - 3, 4 463
AK028027.1 - 3, 4 477
AK049398.1 - 3, 4 449
AK083888.1 - 3, 4 463
AK132029.1 - 3, 4 472
AK145755.1 - 3, 4 454
AK146305.1 - 3, 4 497
AK146473.1 - 3, 4 449
AK150750.1 - 3, 4 471
AK151106.1 - 3, 4 417
AK151147.1 - 3, 4 417
AK151485.1 - 3, 4 474
AK151564.1 - 3, 4 417
AK151640.1 - 3, 4 472
AK151687.1 - 3, 4 445
AK152272.1 - 3, 4 445
AK194194.1 - 2, 3 184
AK198840.1 - 2, 3 184
AK206882.1 - 2, 3 184
BC043084.1 - 3, 4 446 6417343
BC054811.1 - 3, 4 462 6839739
D78188.1 - 3, 4 458
U20290.1 - 3, 4 480

View Full Product Details