Gene Transcripts
- Gene Symbol
- Banf1
- Entrez Gene ID
- 23825
- Gene Name
- barrier to autointegration factor 1
- Gene Aliases
- Baf, Bcrp1, L2bp1/Baf
- Chromosome Location
- Chr. 19: 5364633 - 5366876 on Build GRCm38
- UniGene ID
- Mm.358649
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001038231.2 |
NP_001033320.1
|
2, 3 |
242 |
|
NM_001286608.1 |
NP_001273537.1
|
2, 3 |
584 |
|
NM_011793.3 |
NP_035923.1
|
2, 3 |
450 |
|
GenBank mRNA |
AB025349.1 |
- |
2, 3 |
182 |
|
AF037080.1 |
- |
1, 2 |
125 |
|
AK003025.1 |
- |
2, 3 |
226 |
|
AK004212.1 |
- |
2, 3 |
209 |
|
AK010257.1 |
- |
2, 3 |
228 |
|
AK011079.1 |
- |
2, 3 |
231 |
|
AK011100.1 |
- |
2, 3 |
229 |
|
AK012588.1 |
- |
2, 3 |
212 |
|
AK088896.1 |
- |
2, 3 |
206 |
|
AK189231.1 |
- |
1, 2 |
143 |
|
BC003709.1 |
- |
2, 3 |
219 |
3599828 |
BY111854.1 |
- |
2, 3 |
315 |
|
CK780850.1 |
- |
2, 3 |
214 |
|
View Full Product Details