Gene Transcripts

Gene Symbol
PLAC1
Entrez Gene ID
10761
Gene Name
placenta specific 1
Gene Aliases
CT92, OOSP2L
Chromosome Location
Chr. X: 134565838 - 134764322 on Build GRCh38
UniGene ID
Hs.496811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316887.1 NP_001303816.1 4 764
NM_001316888.1 NP_001303817.1 3 792
NM_001316889.1 NP_001303818.1 2 674
NM_021796.3 NP_068568.1 3 746
XM_011531257.2 XP_011529559.1 3 672
XM_011531260.2 XP_011529562.1 3 786
XM_017029236.1 XP_016884725.1 2 714
GenBank mRNA AF234654.1 - 3 751
AK075086.1 - 3 746
BC022335.1 - 3 731 4769552
BC066327.1 - 2 674 30339131

View Full Product Details