Gene Transcripts

Gene Symbol
Arpc5l
Entrez Gene ID
74192
Gene Name
actin related protein 2/3 complex, subunit 5-like
Gene Aliases
2010015J01Rik, AI852867, ARC16-2, AW555592, AW742746
Chromosome Location
Chr. 2: 39005272 - 39015878 on Build GRCm38
UniGene ID
Mm.38155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028809.1 NP_083085.1 4 477
GenBank mRNA AK008255.1 - 4 477
AK037305.1 - 3 3368
BC024482.1 - 4 500 4977391

View Full Product Details