Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100397.1 | NP_001093867.1 | 5 | 607 | |
NM_001282899.1 | NP_001269828.1 | 5 | 478 | ||
NM_001282900.1 | NP_001269829.1 | 4 | 424 | ||
NM_001282901.1 | NP_001269830.1 | 2 | 212 | ||
NM_007023.3 | NP_008954.2 | 8 | 848 | ||
XM_005246246.2 | XP_005246303.1 | 7 | 661 | ||
XM_006712205.3 | XP_006712268.1 | 7 | 666 | ||
XM_011510505.2 | XP_011508807.1 | 7 | 695 | ||
XM_017003196.1 | XP_016858685.1 | 8 | 715 | ||
XM_017003197.1 | XP_016858686.1 | 4 | 364 | ||
GenBank mRNA | AB027471.1 | - | 8 | 718 | |
AB209681.1 | - | 8 | 777 | ||
AK294433.1 | - | 2 | 136 | ||
AK294445.1 | - | 5 | 327 | ||
AK294914.1 | - | 2 | 165 | ||
AK294997.1 | - | 8 | 813 | ||
AK295002.1 | - | 4 | 359 | ||
AK296340.1 | - | 4 | 419 | ||
AK302732.1 | - | 3 | 203 | ||
AK304278.1 | - | 5 | 367 | ||
AK311165.1 | - | 4 | 246 | ||
AK311500.1 | - | 5 | 607 | ||
AK311575.1 | - | 3 | 266 | ||
AK311697.1 | - | 5 | 584 | ||
AK313084.1 | - | 8 | 717 | ||
AK316014.1 | - | 5 | 442 | ||
AK316072.1 | - | 5 | 374 | ||
BC024004.2 | - | 8 | 687 | 4823935 | |
BC040183.2 | - | 5 | 608 | 4801092 | |
DC335305.1 | - | 5 | 478 | ||
KJ534924.1 | - | 8 | 625 | ||
KJ534925.1 | - | 7 | 607 | ||
KJ534978.1 | - | 8 | 661 | ||
U78516.1 | - | 8 | 769 |