Gene Transcripts

Gene Symbol
Atox1
Entrez Gene ID
84355
Gene Name
antioxidant 1 copper chaperone
Gene Aliases
Atx1
Chromosome Location
Chr. 10: 40790850 - 40805886 on Build Rnor_6.0
UniGene ID
Rn.11992
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053359.2 NP_445811.1 4 465
GenBank mRNA AF127137.1 - 4 401
AF177671.1 - 4 480
BC058458.1 - 4 489 6919265
FQ220104.1 - 4 466

View Full Product Details