Gene Transcripts

Gene Symbol
OPN1SW
Entrez Gene ID
611
Gene Name
opsin 1 (cone pigments), short-wave-sensitive
Gene Aliases
BCP, BOP, CBT
Chromosome Location
Chr. 7: 128772489 - 128775790 on Build GRCh38
UniGene ID
Hs.656404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001708.2 NP_001699.1 4 751
GenBank mRNA DQ822478.1 - 4 751

View Full Product Details