Gene Transcripts
- Gene Symbol
- ANKRD20A2
- Entrez Gene ID
- 441430
- Gene Name
- ankyrin repeat domain 20 family member A2
- Gene Aliases
- Chromosome Location
- Chr. 9: 40223285 - 40282360 on Build GRCh38
- UniGene ID
- Hs.632663
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A1
- Entrez Gene ID
- 84210
- Gene Name
- ankyrin repeat domain 20 family member A1
- Gene Aliases
- ANKRD20A
- Chromosome Location
- Chr. 9: 67859315 - 67902394 on Build GRCh38
- UniGene ID
- Hs.632671
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A3
- Entrez Gene ID
- 441425
- Gene Name
- ankyrin repeat domain 20 family member A3
- Gene Aliases
- Chromosome Location
- Chr. 9: 66093946 - 66153193 on Build GRCh38
- UniGene ID
- Hs.632663
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A4
- Entrez Gene ID
- 728747
- Gene Name
- ankyrin repeat domain 20 family member A4
- Gene Aliases
- Chromosome Location
- Chr. 9: 64369563 - 64412691 on Build GRCh38
- UniGene ID
- Hs.645502
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A5P
- Entrez Gene ID
- 440482
- Gene Name
- ankyrin repeat domain 20 family member A5, pseudogene
- Gene Aliases
- ANKRD20A5
- Chromosome Location
- Chr. 18: 14179097 - 14227050 on Build GRCh38
- UniGene ID
- Hs.632352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040113.1 |
|
13, 14 |
1607 |
|
- Gene Symbol
- LOC105379426
- Entrez Gene ID
- 105379426
- Gene Name
- ankyrin repeat domain-containing protein 20A2
- Gene Aliases
- Chromosome Location
- Chr. 3|NT_167215: 86 - 16759 on Build GRCh38
- UniGene ID
- Hs.679496
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016022.1 |
- |
10, 11 |
1003 |
|
View Full Product Details