Gene Transcripts

Gene Symbol
LAT2
Entrez Gene ID
7462
Gene Name
linker for activation of T-cells family member 2
Gene Aliases
HSPC046, LAB, NTAL, WBSCR15, WBSCR5, WSCR5
Chromosome Location
Chr. 7: 74209757 - 74229834 on Build GRCh38
UniGene ID
Hs.741249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014146.3 NP_054865.2 5, 6 599
NM_032463.2 NP_115852.1 6, 7 718
NM_032464.2 NP_115853.2 6, 7 788
XM_011516558.2 XP_011514860.1 6, 7 628
GenBank mRNA AF252611.1 - 6, 7 539
AF252612.1 - 5, 6 350
AF252613.1 - 5, 6 335
AF252614.1 - 6, 7 469
AF257135.1 - 4, 5 270
AK002099.1 - 6, 7 578
AK092904.1 - 6, 7 929
AK290074.1 - 6, 7 466
AK290916.1 - 5, 6 347
AY190023.1 - 4, 5 280
BC001609.1 - 6, 7 600 3543310
BC009204.2 - 4, 5 272 4054137

View Full Product Details