Gene Transcripts

Gene Symbol
MCEE
Entrez Gene ID
84693
Gene Name
methylmalonyl-CoA epimerase
Gene Aliases
GLOD2
Chromosome Location
Chr. 2: 71109676 - 71130288 on Build GRCh38
UniGene ID
Hs.94949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032601.3 NP_115990.3 2 331
GenBank mRNA AF364547.1 - 2 304
BC020825.1 - 2 296 4734422
BM271757.1 - 2 353

View Full Product Details