Gene Transcripts
- Gene Symbol
- LINC00696
- Entrez Gene ID
- 100128378
- Gene Name
- long intergenic non-protein coding RNA 696
- Gene Aliases
- C3orf74
- Chromosome Location
- Chr. 3: 52062094 - 52065112 on Build GRCh38
- UniGene ID
- Hs.640106
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027331.1 |
|
1 |
2852 |
|
View Full Product Details