Gene Transcripts

Gene Symbol
Dupd1
Entrez Gene ID
361003
Gene Name
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases
Chromosome Location
Chr. 15: 2766929 - 2806556 on Build Rnor_6.0
UniGene ID
Rn.162035
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108368.1 NP_001101838.1 4 1062
XM_006251647.2 XP_006251709.1 4 1006

View Full Product Details