Gene Transcripts

Gene Symbol
Krtap16-5
Entrez Gene ID
680703
Gene Name
keratin associated protein 16-5
Gene Aliases
Chromosome Location
Chr. 11: 28899792 - 28900376 on Build Rnor_6.0
UniGene ID
Rn.218556
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109424.1 NP_001102894.1 1 404

View Full Product Details